Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrinogen gamma chain Antibody (5A6), Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | Fibrinogen gamma chain |
---|---|
Clone | 5A6 |
Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
Fibrinogen gamma chain Monoclonal specifically detects Fibrinogen gamma chain in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Fibrinogen gamma chain | |
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
Monoclonal | |
Ascites | |
RUO | |
Human | |
fibrinogen gamma chain, fibrinogen, gamma polypeptide | |
FGG | |
IgG2b | |
Unpurified |
5A6 | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer | |
Ascites with 0.03% Sodium Azide | |
2266 | |
Purified recombinant fragment of human FGG expressed in E. Coli. | |
Primary | |
52 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title