Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrinogen gamma chain Antibody (OTI2D2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270732
Description
Fibrinogen gamma chain Monoclonal specifically detects Fibrinogen gamma chain in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Fibrinogen gamma chain | |
Monoclonal | |
LYOPH | |
Western Blot 1:400-4000, Immunohistochemistry 1:150 | |
fibrinogen gamma chain, fibrinogen, gamma polypeptide | |
Mouse | |
46.4 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2D2 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
FGG | |
Full length human recombinant protein of human FGG(NP_000500) produced in HEK293T cell. | |
Affinity Purified | |
Angiogenesis, Cancer | |
2266 | |
Human, Mouse, Monkey | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction