Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrinogen gamma chain Mouse anti-Human, Clone: 2B2B8, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 661581IG150UL
Description
The protein encoded by this gene is the gamma component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia and thrombophilia. Alternative splicing results in two transcript variants encoding different isoforms.Specifications
| Fibrinogen gamma chain | |
| Monoclonal | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02679 | |
| FGG | |
| Fibrinogen gamma chain Fusion Protein Ag8817 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 2B2B8 | |
| Unconjugated | |
| FGG | |
| FGG, fibrinogen gamma chain | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2266 | |
| -20°C | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction