Learn More
Invitrogen™ Fibrinogen gamma Monoclonal Antibody (5A6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515902
Description
MA5-15902 targets FGG in indirect ELISA, IF and WB applications and shows reactivity with Human samples. The MA5-15902 immunogen is purified recombinant fragment of human FGG expressed in E. Coli. . MA5-15902 detects FGG which has a predicted molecular weight of approximately 52kDa.
The protein encoded by this gene is the gamma component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia and thrombophilia. Alternative splicing results in two transcript variants encoding different isoforms.
Specifications
Fibrinogen gamma | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.03% sodium azide | |
P02679, Q8VCM7 | |
Fgg | |
Purified recombinant fragment of human FGG expressed in E. Coli. | |
RUO | |
2266, 99571 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
5A6 | |
Unconjugated | |
Fgg | |
3010002H13Rik; AI256424; FGG; fibg; fibrinogen; fibrinogen gamma chain; Fibrinogen gamma-B chain; fibrinogen gamma-chain; fibrinogen, gamma polypeptide; gamma'; gamma-fibrinogen; PRO2061; testicular tissue protein Li 70; Unknown (protein for MGC:127951) | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.