Learn More
Fibrinogen Goat anti-Mouse, Polyclonal, Nordic

Goat Polyclonal Antibody
Supplier: Vector Laboratories GAM/FBG/7S

Description
Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance of the antiserum. Diluted antiserum should be stored at 4 C, not refrozen, and preferably used the same day. In immunoelectrophoresis use 2 ul serum or equivalent against 120 ul antiserum. In double radial immunodiffusion (Ouchterlony) use a rosette arrangement with 10 ul antiserum in 3 mm diameter center well and 2 ul serum samples (neat and serially diluted in 2 mm diameter peripheral wells. Different bleedings of the immunized animals are pooled to obtain a broad spectrum balanced against the varying concentrations of the individual serum protein components.
Fibrinogen Polyclonal antibody specifically detects Fibrinogen in Mouse samples. It is validated for Dot Blot, ELISA, Immunoblot, Immunocytochemistry, ImmunofluorescenceSpecifications
Fibrinogen | |
Polyclonal | |
Unconjugated | |
FGG | |
2510049G14Rik; 3010002H13Rik; A-alpha fibrinogen; Ab1-181; Ab1-216; Ac1-581; Ac1873; AI256424; ENSMUSG00000059807; epididymis secretory sperm binding protein Li 78p; Fba5e; Fga; FGB; Fgg; Fib; Fib2; fibg; fibrinogen; fibrinogen A-alpha chain; fibrinogen alpha chain; Fibrinogen beta chain; fibrinogen gamma chain; Fibrinogen gamma-B chain; fibrinogen gamma-chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; fibrinogen, B beta polypeptide; fibrinogen, beta polypeptide; fibrinogen, gamma polypeptide; Fibrinopeptide A; Fibrinopeptide B; Gamma'; gamma-fibrinogen; HEL-S-78p; liver regeneration-related protein LRRG036/LRRG043/LRRG189; mgC119422; mgC119423; mgC119425; PRO2061; testicular tissue protein Li 70; Unknown (protein for mgC:127951) | |
Mouse fibrinogen | |
10 mg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Dot Blot, ELISA, Immunoblot, Immunocytochemistry, Immunofluorescence | |
10 mg/mL | |
PBS with no preservative; pH | |
E9PV24, Q8K0E8, Q8VCM7 | |
Goat | |
Protein G | |
RUO | |
110135, 14161, 99571 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.