Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fibrinogen Monoclonal Antibody (27C8)
GREENER_CHOICE

Catalog No. PIMA120039
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA120039 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA120039 Supplier Invitrogen™ Supplier No. MA120039
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-20039 detects fibrinogen in human samples.

Fibrinogen, the precursor of fibrin, is the coagulable protein in the blood plasma of vertebrates. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. Fibrinogen consists as a dimer of 3 pairs of non-identical chains FGA, FGB, and FGG that are cross-linked by disulfide bonds in their N-terminal segments. The molecule has 2 terminal D domains and one central E domain, all three domains are separated when fibrinogen is degradated by plasmin. Mutations in this gene lead to several disorders, including dysfibrinogenemia, afibrinogenemia, and renal amyloidosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrinogen
Applications ELISA, Western Blot
Classification Monoclonal
Clone 27C8
Concentration 3.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FGB
Gene Accession No. P02671, P02675, P02679
Gene Alias 2510049G14Rik; 3010002H13Rik; A-alpha fibrinogen; Ab1-181; Ab1-216; Ac1-581; Ac1873; AI256424; carboxy-terminal region; ENSMUSG00000059807; epididymis secretory sperm binding protein Li 78p; Fba5e; Fga; Fgb; Fgg; Fib; Fib2; fibg; fibrinogen; fibrinogen A-alpha chain; fibrinogen A-alpha-chain; fibrinogen alpha chain; fibrinogen alpha-E subunit; fibrinogen beta chain; Fibrinogen gamma chain; fibrinogen gamma polypeptide; fibrinogen gamma-B chain; fibrinogen gamma-chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; fibrinogen, B beta polypeptide; fibrinogen, beta polypeptide; fibrinogen, gamma polypeptide; Fibrinopeptide A; Fibrinopeptide B; Gamma'; gamma-fibrinogen; HEL-S-78p; Liver regeneration-related protein LRRG036/LRRG043/LRRG189; MGC119422; MGC119423; MGC119425; PRO2061; testicular tissue protein Li 70; Unknown (protein for MGC:127951)
Gene Symbols FGA, FGB, FGG
Host Species Mouse
Immunogen Fibrin degradation products.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2243, 2244, 2266
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.