Learn More
Invitrogen™ Fibrinogen Polyclonal Antibody
Description
Sheep anti Human fibrinogen antibody recognizes human fibrinogen, a complex approximately 340 kDa hetero-hexameric (di-trimeric) glycoprotein consisting of 3 pairs of alpha, beta, and gamma chains linked by a series of 29 disulphide bonds (Henschen et al. 1983).
Specifications
Specifications
| Antigen | Fibrinogen |
| Applications | ELISA, Immunohistochemistry (Frozen), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Glycine buffered saline with <0.1% EACA, <0.01% benzamidine, <1mM EDTA and <0.1% sodium azide; pH 7.4 |
| Gene | FGB |
| Gene Accession No. | E9PV24, P02671, P02675, P02679, P02680, P06399, P14480, Q8K0E8, Q8VCM7 |
| Gene Alias | 2510049G14Rik; 3010002H13Rik; A-alpha fibrinogen; Ab1-181; Ab1-216; Ac1-581; Ac1873; AI256424; carboxy-terminal region; ENSMUSG00000059807; epididymis secretory sperm binding protein Li 78p; Fba5e; Fga; Fgb; Fgg; Fib; Fib2; fibg; fibrinogen; fibrinogen A-alpha chain; fibrinogen A-alpha-chain; fibrinogen alpha chain; fibrinogen alpha-E subunit; fibrinogen beta chain; Fibrinogen gamma chain; fibrinogen gamma polypeptide; fibrinogen gamma-B chain; fibrinogen gamma-chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; fibrinogen, B beta polypeptide; fibrinogen, beta polypeptide; fibrinogen, gamma polypeptide; Fibrinopeptide A; Fibrinopeptide B; Gamma'; gamma-fibrinogen; HEL-S-78p; Liver regeneration-related protein LRRG036/LRRG043/LRRG189; MGC119422; MGC119423; MGC119425; PRO2061; testicular tissue protein Li 70; Unknown (protein for MGC:127951) |
| Gene Symbols | FGA, FGB, FGG |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.