Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrinogen Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $437.00
Specifications
Antigen | Fibrinogen |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145489
|
Invitrogen™
PA5145489 |
200 μL |
Each for $437.00
|
|
|||||
FEPA5145490
|
Invitrogen™
PA5145490 |
20 μL |
Each for $191.80
|
|
|||||
Description
Fibrinogen, the precursor of fibrin, is the coagulable protein in the blood plasma of vertebrates. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. Fibrinogen consists as a dimer of 3 pairs of non-identical chains FGA, FGB, and FGG that are cross-linked by disulfide bonds in their N-terminal segments. The molecule has 2 terminal D domains and one central E domain, all three domains are separated when fibrinogen is degradated by plasmin. Mutations in this gene lead to several disorders, including dysfibrinogenemia, afibrinogenemia, and renal amyloidosis.Specifications
Fibrinogen | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Pig | |
P14460, P14477 | |
100514354, 100626178, 403164 | |
Native Pig FG protein (Accession #P14460 & P14477). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
2510049G14Rik; 3010002H13Rik; A-alpha fibrinogen; Ab1-181; Ab1-216; Ac1-581; Ac1873; AI256424; carboxy-terminal region; ENSMUSG00000059807; epididymis secretory sperm binding protein Li 78p; Fba5e; Fga; Fgb; Fgg; Fib; Fib2; fibg; fibrinogen; fibrinogen A-alpha chain; fibrinogen A-alpha-chain; fibrinogen alpha chain; fibrinogen alpha-E subunit; fibrinogen beta chain; Fibrinogen gamma chain; fibrinogen gamma polypeptide; fibrinogen gamma-B chain; fibrinogen gamma-chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; fibrinogen, B beta polypeptide; fibrinogen, beta polypeptide; fibrinogen, gamma polypeptide; Fibrinopeptide A; Fibrinopeptide B; Gamma'; gamma-fibrinogen; HEL-S-78p; Liver regeneration-related protein LRRG036/LRRG043/LRRG189; MGC119422; MGC119423; MGC119425; PRO2061; testicular tissue protein Li 70; Unknown (protein for MGC:127951) | |
FGA, FGB, FGG | |
IgG | |
Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title