Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FibrinogenGoat anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501989990
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9990 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9990 Supplier Rockland Immunochemicals Supplier No. 100101240
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-goat serum, purified and partially purified Fibrinogen [Human Plasma]. Cross reactivity against Fibrinogen from other sources is unknown.

Fibrinogen, the precursor of fibrin, is the coagulable protein in the blood plasma of vertebrates. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. Fibrinogen consists as a dimer of 3 pairs of non-identical chains FGA, FGB, and FGG that are cross-linked by disulfide bonds in their N-terminal segments. The molecule has 2 terminal D domains and one central E domain, all three domains are separated when fibrinogen is degradated by plasmin. Mutations in this gene lead to several disorders, including dysfibrinogenemia, afibrinogenemia, and renal amyloidosis.

Specifications

Antigen Fibrinogen
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 90 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene FGB
Gene Accession No. P02671, P02675, P02679
Gene Alias 2510049G14Rik; 3010002H13Rik; Ab1-181; Ab1-216; Ac1-581; Ac1873; AI256424; ENSMUSG00000059807; epididymis secretory sperm binding protein Li 78p; Fba5e; FGA; Fgb; FGG; Fib; Fib2; Fibrinogen alpha chain; Fibrinogen beta chain; Fibrinogen gamma chain; fibrinogen gamma-chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; fibrinogen, B beta polypeptide; fibrinogen, beta polypeptide; fibrinogen, gamma polypeptide; Fibrinopeptide A; Fibrinopeptide B; gamma-fibrinogen; HEL-S-78p; Liver regeneration-related protein LRRG036/LRRG043/LRRG189; MGC119422; MGC119423; MGC119425; PRO2061; testicular tissue protein Li 70
Gene Symbols FGA, FGB, FGG
Host Species Goat
Immunogen Fibrinogen [Human Plasma]
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2243, 2244, 2266
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.