Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibroblast Activation Protein alpha/FAP Antibody (983802), Janelia Fluor™ 646, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB9727J
This item is not returnable.
View return policy
Description
Fibroblast Activation Protein alpha/FAP Monoclonal antibody specifically detects Fibroblast Activation Protein alpha/FAP in Mouse samples. It is validated for Flow Cytometry, ELISASpecifications
Fibroblast Activation Protein alpha/FAP | |
Monoclonal | |
Janelia Fluor 646 | |
170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin | |
Mouse myeloma cell line NS0-derived recombinant mouse Fibroblast Activation Protein alpha/FAP, Leu26-Asp761, Accession # P97321 | |
0.1 mL | |
Cancer, Protein Phosphatase | |
2191 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, ELISA | |
983802 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction