Learn More
Fibroblast Activation Protein Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $649.44
Specifications
| Antigen | Fibroblast Activation Protein |
|---|---|
| Concentration | 1.1 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1944
![]()
|
Rockland Immunochemicals
600401A73 |
100 μg |
Each for $649.44
|
|
|||||
50-199-1945
![]()
|
Rockland Immunochemicals
600401A73S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is known to react with mouse FAP protein. A BLAST analysis was used to suggest partial cross-reactivity with FAP from human and rat sources based on ∼88% homology with the immunizing sequence. Reactivity with FAP from other sources has not been determined.
The protein encoded by this gene is a homodimeric integral membrane gelatinase belonging to the serine protease family. It is selectively expressed in reactive stromal fibroblasts of epithelial cancers, granulation tissue of healing wounds, and malignant cells of bone and soft tissue sarcomas. This protein is thought to be involved in the control of fibroblast growth or epithelial-mesenchymal interactions during development, tissue repair, and epithelial carcinogenesis.Specifications
| Fibroblast Activation Protein | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| P97321 | |
| 14089 | |
| Primary | |
| Antibody |
| 1.1 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| 170 kDa melanoma membrane-bound gelatinase; Antiplasmin-cleaving enzyme FAP, soluble form; APCE; Dipeptidyl peptidase FAP; DPPIV; FAP; FAPA; FAPalpha; fibroblast activation protein; Fibroblast activation protein alpha; fibroblast activation protein, alpha; Gelatine degradation protease FAP; Integral membrane serine protease; Post-proline cleaving enzyme; Prolyl endopeptidase FAP; Seprase; Serine integral membrane protease; SIMP; Surface-expressed protease | |
| FAP | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| FAP |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.