Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fibroblasts/Epithelial Cells Monoclonal Antibody (D7-FIB)
GREENER_CHOICE

Catalog No. PIMA528455
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528455 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528455 Supplier Invitrogen™ Supplier No. MA528455
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes approximately a 112kDa molecule expressed on the cell surface of human fibroblasts. The antigen is not expressed by peripheral blood cells, and is found at low levels on a minority of melanoma cell lines.

Fibroblasts/Epithelial Cells antibody recognizes an approximately 112 kDa molecule expressed on the cell surface of human fibroblasts. The antigen is not expressed by peripheral blood cells, and is found at low levels on a minority of melanoma cell lines. Studies upon the antigen have shown it to be sensitive to SDS, but resistant to trypsin, tunicamycin and collagenase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibroblasts/Epithelial Cells
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone D7-FIB
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Alias fibro-blasts
Host Species Mouse
Immunogen Human foreskin fibroblasts.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.