Learn More
Invitrogen™ Fibroblasts Monoclonal Antibody (D7-FIB), PE

Description
This antibody does not react with mouse or rat. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human Fibroblasts/Epithelial Cells antibody, clone D7-FIB recognizes a approximately 112kDa molecule expressed on the cell surface of human fibroblasts.
Specifications
Specifications
| Antigen | Fibroblasts |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | D7-FIB |
| Concentration | Conc. Not Determined |
| Conjugate | PE |
| Formulation | PBS with 1% BSA, 5% sucrose and 0.09% sodium azide |
| Gene Alias | fibro-blasts |
| Host Species | Mouse |
| Immunogen | Human foreskin fibroblasts. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.