Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fibroblasts Monoclonal Antibody (ER-TR7)
GREENER_CHOICE

Catalog No. 01676513 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-513 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-513 Supplier Invitrogen™ Supplier No. MA140076
  • $782.00
    $629.65 / Each of 1

    Save $152.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

MA1-40076 detects Fibroblasts from mouse samples. MA1-40076 is cross reactive with human tonsil fibroblasts. It does not cross react with purified laminin, collagen types I-V, fibronectin, heparin sulfate proteoglycan,entactin or nidogen. MA1-40076 has been successfully used in immunohistochemistry (frozen and parrafin), flow cytometry, and immunofluorescence applications. Suggested positive control is spleen The MA1-40076 immunogen is mouse thymic stromal cells. Suggested positive control: Spleen; Suggested negative control: Lymphoid cells.

Fibroblasts are the least specialized cells in the connective-tissue family. Thay are dispersed in connective tissue throughout the body, where they secrete a nonrigid extracellular matrix that is rich in type I and/or type III collagen. The activation of fibroblasts by inflammatory stimuli results in their migration, proliferation and deposition of extracellular matrix components, important features involved in both wound healing and fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibroblasts
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ER-TR7
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene Alias fibro-blasts
Host Species Rat
Immunogen Mouse thymic stromal cells.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.