Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibromodulin/FMOD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216494
Description
Fibromodulin/FMOD Polyclonal specifically detects Fibromodulin/FMOD in Human samples. It is validated for Western Blot.Specifications
Fibromodulin/FMOD | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
Collagen-binding 59 kDa protein, fibromodulin, fibromodulin proteoglycan, FM, Keratan sulfate proteoglycan fibromodulin, SLRR2EKSPG fibromodulin | |
Rabbit | |
43 kDa | |
0.1 mL | |
Signal Transduction | |
2331 | |
Human | |
IgG |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q06828 | |
FMOD | |
Recombinant protein encompassing a sequence within the center region of human Fibromodulin. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Expected to cross react based on sequence identity: Chicken (80%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction