Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fibromodulin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109733
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109733 Supplier Invitrogen™ Supplier No. PA5109733
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QYEDDPHWWF HYLRSQQSTY YDPYDPYPYE TYEPYPYGVD EGPAYTYGSP SPPDPRDCPQ ECDCPPNFPT AMYCDNRNLK YLPFVPSRMK YVYFQNNQIT SIQEGVFDNA TGLLWIALHG NQITSDKVGR KVFSKLRHLE RLYLDHNNLT RMPGPLPRSL RELHLDHNQI SRVPNNALEG LENLTALYLQ HNEIQEVGSS MRGLRSLILL DLSYNHLRKV PDGLPSALEQ LYMEHNNVYT VPDSYFRGAP KLLYVRLSHN SLTNNGLASN TFNSSSLLEL DLSYNQLQKI PPVNTNLENL YLQGNRINEF SISSFCTVVD VVNFSKLQVL RLDGNEIKRS AMPADAPLCL RLASLIEI.

Fibromodulin is a member of a family of small interstitial proteoglycans, containing a central region composed of leucine-rich repeats with 4 keratan sulfate chains flanked by disulfide-bonded terminal domains. It may participate in the assembly of the extracellular matrix as it interacts with type I and type II collagen fibrils and inhibits fibrillogenesis in vitro. It may also regulate TGF-beta activities by sequestering TGF-beta into the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibromodulin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FMOD
Gene Accession No. P50608, P50609, Q06828
Gene Alias AI131919; AU041740; Collagen-binding 59 kDa protein; fibromodulin; FM; FMOD; Keratan sulfate proteoglycan fibromodulin; KSPG fibromodulin; SLRR2E
Gene Symbols FMOD
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 19-376 of human FMOD (NP_002014.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14264, 2331, 64507
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.