Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibronectin Antibody (014), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289407R
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Western Blot, ELISA.Specifications
Fibronectin | |
Monoclonal | |
DyLight 550 | |
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Fibronectin (Ser 607-Pro 1265; CAD91166). | |
0.1 mL | |
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
2335 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
14 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction