Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibronectin Antibody (2F4), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222113F
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Fibronectin | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
FN1 | |
Purified recombinant fragment of human Fibronectin expressed in E. coli. [UniProt# P02751] | |
0.1 mL | |
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
2335 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) | |
2F4 | |
Flow Cytometry, ELISA, Immunohistochemistry-Paraffin | |
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction