Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibronectin Antibody (568), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP248018APCCY7
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Flow Cytometry.Specifications
Fibronectin | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
FN1 | |
High molecular weight proteins secreted by cultivated human fibroblasts (Uniprot: P02751) | |
0.1 mL | |
Primary | |
Epitope of this monoclonal antibody is located in the 8th type III repeat in the cell-binding region of fibronectin. Fibronectins are disulfide-linked, dimeric glycoproteins of ∼440kDa. They possess at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectins are involved in cell adhesion, tissue organization, and wound healing. Fibronectins are present in basement membranes, interstitial connective tissue matrix, and blood. Cellular fibronectin is widely distributed in the stroma of many malignant tumors. This monoclonal antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
568 | |
Flow Cytometry | |
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
Mouse | |
Protein A or G purified | |
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
2335.0 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction