Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibronectin Antibody (568), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2450180.1MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
Fibronectin | |
Monoclonal | |
0.2mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
P02751, P02751 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Epitope of this MAb is located in the 8th type III repeat in the cell-binding region of fibronectin. Fibronectins are disulfide-linked, dimeric glycoproteins of ∼440kDa. They possess at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectins are involved in cell adhesion, tissue organization, and wound healing. Fibronectins are present in basement membranes, interstitial connective tissue matrix, and blood. Cellular fibronectin is widely distributed in the stroma of many malignant tumors. This MAb is excellent for staining of formalin-fixed, paraffin-embedded tissues. | |
Store at 4C. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
568 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
FN1 | |
High molecular weight proteins secreted by cultivated human fibroblasts | |
0.1 mg | |
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
2335 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction