Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibronectin Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP191258AF594
Description
Fibronectin Polyclonal specifically detects Fibronectin in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Fibronectin | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
Rabbit | |
0.1 mL | |
2335 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
FN1 | |
A synthetic peptide made toward the C-terminal region of the human Fibronectin protein (within residues 2250-2300). [Swiss-Prot: P02751] | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Feline | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction