Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibronectin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB110415620.1ML
Description
Fibronectin Polyclonal specifically detects Fibronectin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Fibronectin | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:250, Immunohistochemistry-Frozen 1:50-1:250 | |
| CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
| Rabbit | |
| 0.1 mL | |
| 2335 | |
| Human | |
| Whole Antisera |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Whole antisera with sodium azide | |
| FN1 | |
| Fibronectin isolated from a pool of normal human plasma. | |
| Primary | |
| This reacts with human fibronectin in liver, tonsil, skin and kidney. Traces of contaminating antibodies have been removed by solid-phase absorption. Only the fibronectin arch was observed in crossed immunoelectrophoresis. | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction