Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibronectin Antibody (OTI5C1), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270762H
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Fibronectin | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
FN1 | |
Recombinant protein fragment corresponding to amino acids 32-307 of human Fibronectin (NP_473375) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI5C1 | |
Western Blot | |
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
Mouse | |
Affinity Purified | |
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
2335.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction