Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin Mouse anti-Mouse, Porcine, Human, MilliporeSigma™

Catalog No. MAB1926I25 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1926-I-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1926-I-25 Supplier MilliporeSigma Supplier No. MAB1926I25UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mouse monoclonal Anti-Fibronectin, cell binding domain, clone P1H11 Antibody.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibronectin
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols FN1,FN
Host Species Mouse
Immunogen Human Plasma Fibronectin.
Purification Method Protein G Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone P1H11 detects fibronectin in human.
Target Species Mouse, Pig, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.