Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin Polyclonal Antibody, Invitrogen™

Catalog No. PIPA596698
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596698 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596698 Supplier Thermo Scientific Supplier No. PA596698

Rabbit Polyclonal Antibody

Immunogen sequence: MLRGPGPGLL LLAVQCLGTA VPSTGASKSK RQAQQMVQPQ SPVAVSQSKP GCYDNGKHYQ INQQWERTYL GNALVCTCYG GSRGFNCESK PEAEETCFDK YTGNTYRVGD TYERPKDSMI WDCTCIGA; Positive Samples: NIH/3T3; Cellular Location: extracellular matrix, extracellular space

Fibronectin is a disulfide-bonded dimer with a molecular weight range of 230-250 kDa. In the extracellular matrix of several connective tissues and vessels, fibronectin is present as an insoluble protein that is extensively cross-linked by interchain disulfide bonds forming high molecular mass polymers. Fibronectin is most abundant during embryonic development and tissue remodeling. Fibronectin is also present at high concentrations as a soluble plasma protein. Fibronectin is organized as a linear series of repeating modules which form domains for interaction with fibronectin itself, other matrix components (e.g. collagen and heparin) and receptors on cells (e.g. integrins). Fibronectin is present in a soluble dimeric form in plasma, and in a dimeric or multimeric form at the cell surface and in extracellular matrix. Fibronectin is involved in cell adhesion and migration processes including embryogenesis, wound healing, blood coagulation, host defense, and metastasis. Fibronectin has been implicated in carcinoma development in lung cancer. Further, Fibronectin expression is increased especially in non-small cell lung carcinoma. The adhesion of lung carcinoma cells to Fibronectin enhances tumorgenecity and confers resistance to apoptosis induced by standard chemotherapeutic agents. The gene encoding Fibronectin has three regions subject to alternative splicing, with the potential to produce 20 different transcript variants. However, the full-length nature of some Fibronectin variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibronectin
Applications Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
Gene FN1
Gene Accession No. P02751, P04937, P11276
Gene Alias Anastellin, CIG, Cold insoluble globulin (CIG), Cold-insoluble globulin, cumulus cell-specific fibronectin 1 transcript, DKFZp686F10164, DKFZp686H0342, DKFZp686I1370, DKFZp686O13149, E330027I09, ED-B, embryo-specific fibronectin 1 transcript, ferritin L subunit, ferritin L-chain, FIBNEC, fibronectin, fibronectin 1, fibronectin ED-A- region, fibronectin ED-B+ region, fibronectin variable region, FINC, FN, FN1, fn-1, FNZ, GFND, GFND2, LETS, LOW QUALITY PROTEIN: fibronectin, Migration stimulating factor (MSF), migration-stimulating factor, MSF, Ugl-Y1, Ugl-Y2, Ugl-Y3
Gene Symbols FN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-128 of human Fibronectin (NP_0020171)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14268, 2335, 25661
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.