Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibronectin Sheep anti-Human, Rat, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody has been used in 8 publications
Supplier: R&D Systems AF1918SP
This item is not returnable.
View return policy
Description
Fibronectin Polyclonal specifically detects Fibronectin in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.Specifications
Fibronectin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FN1 | |
Human plasma-derived Fibronectin | |
25 μg | |
Primary | |
Detects human Fibronectin in direct ELISAs. Detects human and rat Fibronectin in Western blots. | |
Human, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
Sheep | |
Affinity Purified | |
RUO | |
2335 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only