Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibulin 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157911
Description
Fibulin 1 Polyclonal specifically detects Fibulin 1 in Human samples. It is validated for Western Blot.Specifications
| Fibulin 1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FBLN, FIBL1, FIBL-1, fibulin 1, fibulin-1 | |
| Rabbit | |
| 72 kDa | |
| 100 μL | |
| Cancer | |
| 2192 | |
| Reconstitute in 50μL distilled water to a final antibody concentration of 1mg/mL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P23142 | |
| FBLN1 | |
| Synthetic peptides corresponding to FBLN1(fibulin 1) The peptide sequence was selected from the N terminal of FBLN1 (NP_001987). Peptide sequence CCVKSQETGDLDVGGLQETDKIIEVEEEQEDPYLNDRCRGGGPCKQQCRD. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Yeast: 83%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Yeast | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction