Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Fibulin 1 Monoclonal Antibody (CL0337)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524598
Description
Immunogen sequence: CESGIHNCLP DFICQNTLGS FRCRPKLQCK SGFIQDALGN CIDINECLSI SAPCPIGHTC INTEGSYTCQ KNVPNCGRGY HLNEEGTRCV DVDECAPPAE PCGKGHRCVN SPGSFRCECK TGYYFDGISR MCVDVNE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.
Fibulin-1 is a modular glycoprotein component of the elastic extracellular matrix fibers, basement membranes and blood. Fibulin-1 self associates as well as binds to calcium, fibronectin, laminin, nidogen and fibrinogen. These interactions, individually or in combination, may account for the observed association of Fibulin-1 with basement membranes, connective tissue, elastic fibers and fibrin clots. Fibulin-1 expression is stimulated by estrogen in ovarian cancer cell lines and has been suggested as both an agent of metastasis in ovarian cancer cells and an indicator for predicting cancer risk or aggressiveness in ovarian carcinomas. Other studies point to the inhibition of cancer cell motility with increasing exposure to Fibulin-1. The exact function of Fibulin-1 in the cell is unknown.
Specifications
Fibulin 1 | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P23142 | |
Fbln1 | |
Recombinant protein corresponding to Human Fibulin 1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
CL0337 | |
Unconjugated | |
Fbln1 | |
Basement-membrane protein 90; BM-90; BM-90/fibulin extracellular matrix glyroprotein; extracellular matrix; extracellular matrix and blood protein; FBLN; Fbln1; fbln1c; fbln1d; FIBL1; FIBL-1; fibulin 1; fibulin 1C; fibulin 1D; Fibulin1; fibulin-1; fibulin-1 C varible region; fibulin-1 D; LOW QUALITY PROTEIN: fibulin-1; PP213; wu:fc52c06 | |
Mouse | |
Protein A | |
RUO | |
2192 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction