Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fibulin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103841
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103841 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103841 Supplier Invitrogen™ Supplier No. PA5103841
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FBLN1.

Fibulin-1 is a modular glycoprotein component of the elastic extracellular matrix fibers, basement membranes and blood. Fibulin-1 self associates as well as binds to calcium, fibronectin, laminin, nidogen and fibrinogen. These interactions, individually or in combination, may account for the observed association of Fibulin-1 with basement membranes, connective tissue, elastic fibers and fibrin clots. Fibulin-1 expression is stimulated by estrogen in ovarian cancer cell lines and has been suggested as both an agent of metastasis in ovarian cancer cells and an indicator for predicting cancer risk or aggressiveness in ovarian carcinomas. Other studies point to the inhibition of cancer cell motility with increasing exposure to Fibulin-1. The exact function of Fibulin-1 in the cell is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibulin 1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Fbln1
Gene Accession No. P23142, Q08879
Gene Alias Basement-membrane protein 90; BM-90; BM-90/fibulin extracellular matrix glyroprotein; extracellular matrix; extracellular matrix and blood protein; FBLN; Fbln1; fbln1c; fbln1d; FIBL1; FIBL-1; fibulin 1; fibulin 1C; fibulin 1D; Fibulin1; fibulin-1; fibulin-1 C varible region; fibulin-1 D; LOW QUALITY PROTEIN: fibulin-1; PP213; wu:fc52c06
Gene Symbols Fbln1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FBLN1(Accession P23142), corresponding to amino acid residues S316-P366.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14114, 2192, 315191
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.