Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibulin-2 Polyclonal Antibody, Abbomax

Catalog No. 502203914
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3914 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3914 Supplier Abbomax Inc Supplier No. 600630
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Mouse kidney Cellular location: Extracellular Matrix.

Fibulin-2 Polyclonal antibody specifically detects Fibulin-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Fibulin-2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene FBLN2
Gene Accession No. P37889, P98095
Gene Alias 5730577E14Rik; FBLN2; FIBL2; FIBL-2; fibulin 2; Fibulin2; fibulin-2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the 1140-1165aa of Fibulin-2 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14115, 2199, 282583
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.