Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fibulin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520761
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520761 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520761 Supplier Invitrogen™ Supplier No. PA520761
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is Hela cell lysate. PA5-20761 can be used with blocking peptide PEP-0875.

This gene encodes a member of the fibulin family of extracellular matrix glycoproteins. Like all members of this family, the encoded protein contains tandemly repeated epidermal growth factor-like repeats followed by a C-terminus fibulin-type domain. This gene is upregulated in malignant gliomas and may play a role in the aggressive nature of these tumors. Mutations in this gene are associated with Doyne honeycomb retinal dystrophy. Alternatively spliced transcript variants that encode the same protein have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibulin 3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene EFEMP1
Gene Accession No. Q12805
Gene Alias DHRD; DRAD; EFEMP1; EGF containing fibulin like extracellular matrix protein 1; EGF containing fibulin-like extracellular matrix protein 1; EGF-containing fibulin-like extracellular matrix protein 1; epidermal growth factor-containing fibulin-like extracellular matrix protein 1; extracellular protein S1-5; FBLN3; FBNL; FIBL-3; Fibrillin-like protein; Fibulin-3; FLJ35535; MGC111353; MLVT; MTLV; S1-5; T16
Gene Symbols EFEMP1
Host Species Rabbit
Immunogen A 19 amino acid synthetic peptide near the carboxy terminus of human Fibulin 3 (aa 410 - 460).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2202
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.