Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibulin 5 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2A5C8, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Fibulin 5 |
---|---|
Clone | 2A5C8 |
Concentration | 1.2 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6231
![]() ![]() |
Proteintech Group Inc
600811IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Essential for elastic fiber formation, is involved in the assembly of continuous elastin (ELN) polymer and promotes the interaction of microfibrils and ELN. Stabilizes and organizes elastic fibers in the skin, lung and vasculature. Promotes adhesion of endothelial cells through interaction of integrins and the RGD motif. Vascular ligand for integrin receptors which may play a role in vascular development and remodeling.Specifications
Fibulin 5 | |
1.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ARMD3, DANCE, EVEC, FBLN5, FIBL 5, fibulin 5, UP50, Urine p50 protein | |
Fbln5 | |
IgG1 | |
Caprylic Acid Precipitation | |
Antibody |
2A5C8 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Pig, Rat | |
Q9UBX5, Q9WVH8, Q9WVH9 | |
100155248, 10516, 23876, 29158 | |
Fibulin 5 Fusion Protein Ag2859 | |
Primary | |
-20°C | |
FBLN5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title