Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Fibulin 5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581900
Description
Immunogen sequence: QAQCTNGFDL DRQSGQCLDI DECRTIPEAC RGDMMCVNQN GGYLCIPRTN PVYRGPYSNP YSTPYSGPYP AAAPPLSAPN YPTISRPLIC RFGYQMDESN QCVDVDECAT DSHQCNPTQI CINTEGGYTC SCTDGYWLLE GQCLDIDEC.
Essential for elastic fiber formation, is involved in the assembly of continuous elastin (ELN) polymer and promotes the interaction of microfibrils and ELN. Stabilizes and organizes elastic fibers in the skin, lung and vasculature. Promotes adhesion of endothelial cells through interaction of integrins and the RGD motif. Vascular ligand for integrin receptors which may play a role in vascular development and remodeling.
Specifications
Fibulin 5 | |
Polyclonal | |
Unconjugated | |
FBLN5 | |
A55; ADCL2; ARCL1A; ARMD3; Dance; developmental arteries and neural crest EGF-like protein; Embryonic vascular EGF repeat-containing protein; EVEC; Fbln5; FIBL-5; fibulin 5; fibulin-5; FLJ90059; HNARMD; testis tissue sperm-binding protein Li 75n; UNQ184/PRO210; UP50; urine p50 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10516 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UBX5 | |
FBLN5 | |
Recombinant protein corresponding to Human FBLN5. Recombinant protein control fragment (Product #RP-88946). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction