Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fibulin 5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106664
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106664 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106664 Supplier Invitrogen™ Supplier No. PA5106664
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FBLN5.

Essential for elastic fiber formation, is involved in the assembly of continuous elastin (ELN) polymer and promotes the interaction of microfibrils and ELN. Stabilizes and organizes elastic fibers in the skin, lung and vasculature. Promotes adhesion of endothelial cells through interaction of integrins and the RGD motif. Vascular ligand for integrin receptors which may play a role in vascular development and remodeling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibulin 5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FBLN5
Gene Accession No. Q9UBX5, Q9WVH8, Q9WVH9
Gene Alias A55; ADCL2; ARCL1A; ARMD3; Dance; developmental arteries and neural crest EGF-like protein; Embryonic vascular EGF repeat-containing protein; EVEC; Fbln5; FIBL-5; fibulin 5; fibulin-5; FLJ90059; HNARMD; testis tissue sperm-binding protein Li 75n; UNQ184/PRO210; UP50; urine p50 protein
Gene Symbols FBLN5
Host Species Rabbit
Immunogen A synthesized peptide derived from human FBLN5(Accession Q9UBX5), corresponding to amino acid residues E388-R438.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10516, 23876, 29158
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.