Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Fibulin 5 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518568
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Essential for elastic fiber formation, is involved in the assembly of continuous elastin (ELN) polymer and promotes the interaction of microfibrils and ELN. Stabilizes and organizes elastic fibers in the skin, lung and vasculature. Promotes adhesion of endothelial cells through interaction of integrins and the RGD motif. Vascular ligand for integrin receptors which may play a role in vascular development and remodeling.
Specifications
| Fibulin 5 | |
| Polyclonal | |
| Unconjugated | |
| FBLN5 | |
| A55; ADCL2; ARCL1A; ARMD3; Dance; developmental arteries and neural crest EGF-like protein; Embryonic vascular EGF repeat-containing protein; EVEC; Fbln5; FIBL-5; fibulin 5; fibulin-5; FLJ90059; HNARMD; testis tissue sperm-binding protein Li 75n; UNQ184/PRO210; UP50; urine p50 protein | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10516 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9UBX5 | |
| FBLN5 | |
| Synthetic peptide sequence (RPIKGPREIQLDLE) corresponding to the internal amino acids of FBLN5 (aa 408-422). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction