Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Fibulin 7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557106
Description
Immunogen sequence: ASQNCLSKQQ LLSAIRQLQQ LLKGQETRFA EGIRHMKSRL AALQNSVGRV GPDALPVSCP ALNTPADGRK FGSKYLVDHE Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 75%.
An adhesion molecule that interacts with extracellular matrix molecules in developing teeth and may play important roles in differentiation and maintenance of odontoblasts as well as in dentin formation.
Specifications
| Fibulin 7 | |
| Polyclonal | |
| Unconjugated | |
| FBLN7 | |
| 1600015H20Rik; AI464334; extracellular matrix protein TM14; Fbln7; FIBL-7; fibulin 7; fibulin-7; Protein TM14; RGD1310131; TM14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 129804 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53RD9 | |
| FBLN7 | |
| Recombinant protein corresponding to Human Fibulin 7. Recombinant protein control fragment (Product #RP-96117). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction