Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fibulin 7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532125
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532125 Supplier Invitrogen™ Supplier No. PA532125
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HCT116, MCF-7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

An adhesion molecule that interacts with extracellular matrix molecules in developing teeth and may play important roles in differentiation and maintenance of odontoblasts as well as in dentin formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibulin 7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene FBLN7
Gene Accession No. Q53RD9
Gene Alias 1600015H20Rik; AI464334; extracellular matrix protein TM14; Fbln7; FIBL-7; fibulin 7; fibulin-7; Protein TM14; RGD1310131; TM14
Gene Symbols FBLN7
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Fibulin 7. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129804
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.