Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FICD Monoclonal Antibody (OTI2A12), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA802842
Description
The function remains known.Specifications
| FICD | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BVA6 | |
| FICD | |
| Human recombit protein fragment corresponding to amino acids 61-265 of human FICD produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2A12 | |
| Unconjugated | |
| FICD | |
| HIP13, HYPE, UNQ3041 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11153 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction