Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FICD Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542933
Description
Peptide sequence: GDVRPFIRFI AKCTETTLDT LLFATTEYSV ALPEAQPNHS GFKETLPVKP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
Protein that can both mediate the addition of adenosine 5'-monophosphate (AMP) to specific residues of target proteins (AMPylation), and the removal of the same modification from target proteins (de-AMPylation), depending on the context. The side chain of Glu-231 determines which of the two opposing activities (AMPylase or de-AMPylase) will take place. Acts as a key regulator of the ERN1/IRE1-mediated unfolded protein response (UPR) by mediating AMPylation or de-AMPylation of HSPA5/BiP (PubMed:25601083). In unstressed cells, acts as an adenylyltransferase by mediating AMPylation of HSPA5/BiP at 'Thr-518', thereby inactivating it. In response to endoplasmic reticulum stress, acts as a phosphodiesterase by mediating removal of ATP (de-AMPylation) from HSPA5/BiP at 'Thr-518', leading to restore HSPA5/BiP activity. Although it is able to AMPylate RhoA, Rac and Cdc42 Rho GTPases in vitro, Rho GTPases do not constitute physiological substrates (PubMed:19362538, PubMed:25601083). [UniProt].
Specifications
| FICD | |
| Polyclonal | |
| Unconjugated | |
| Ficd | |
| adenosine monophosphate-protein transferase FICD; AMPylator FICD; D5Ertd40e; De-AMPylase FICD; FIC domain containing; FIC domain protein adenylyltransferase; FIC domain-containing protein; fic S-phase protein cell division homolog; FICD; HIP13; HIP-13; huntingtin interacting protein 13; huntingtin interacting protein E; huntingtin interactor protein E; huntingtin yeast partner E; Huntingtin-interacting protein 13; huntingtin-interacting protein E; Hype; MGC5623; Protein adenylyltransferase FICD; protein adenylyltransferase FICD; adenosine monophosphate-protein transferase FICD; UNQ3041; UNQ3041/PRO9857 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11153 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BVA6 | |
| Ficd | |
| Synthetic peptide directed towards the C-terminal of human FICD (aa 409-458). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction