Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ficolin-2 Antibody (19), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105042H
Description
Ficolin-2 Monoclonal specifically detects Ficolin-2 in Human samples. It is validated for Western Blot, ELISA.Specifications
Ficolin-2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
FCN2 | |
Purified procaryot expressed recombinant human L-Ficolin and full-length L-Ficolin (Ficolin-2) protein | |
0.1 mL | |
Primary | |
Reacts specifically with L-Ficolin. No cross reactivity is seen with M-Ficolin (Ficolin-1) and H-Ficolin (Ficolin-3). | |
Store at 4C in the dark. | |
IgG2a κ |
Western Blot, ELISA | |
19 | |
Western Blot, ELISA | |
37 kDa elastin-binding protein, Collagen/fibrinogen domain-containing protein 2, EBP-37ficolin (collagen/fibrinogen domain-containing lectin) 2 (hucolin), FCNLficolin B, ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin), ficolin-2, Ficolin-B, Ficolin-beta, Hucolin, L-ficolin, P35, Serum lectin p35 | |
Mouse | |
Protein A purified | |
RUO | |
2220 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction