Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ficolin-2 Mouse anti-Human, Clone: 16, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
Antigen | Ficolin-2 |
---|---|
Clone | 16 |
Concentration | 1 mg/ml |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Classification | Monoclonal |
Description
Ficolin-2 Monoclonal specifically detects Ficolin-2 in Human samples. It is validated for Western Blot, ELISA.Specifications
Ficolin-2 | |
1 mg/ml | |
Monoclonal | |
Purified | |
RUO | |
37 kDa elastin-binding protein, Collagen/fibrinogen domain-containing protein 2, EBP-37ficolin (collagen/fibrinogen domain-containing lectin) 2 (hucolin), FCNLficolin B, ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin), ficolin-2, Ficolin-B, Ficolin-beta, Hucolin, L-ficolin, P35, Serum lectin p35 | |
Purified procaryot expressed recombinant human L-Ficolin and full-length L-Ficolin (Ficolin-2) protein adsorbed onto aluminum hydroxide gel | |
Primary | |
Store at 4°C in the dark. |
16 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Unconjugated | |
Mouse | |
10mM Phosphate (pH 7.4) and 0.5M NaCl | |
2220 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title