Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ficolin-3 Antibody (FCN334), Alexa Fluor™ 405, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP223573AF405

Catalog No. NP22373A40


Only null left
Add to Cart

Description

Description

Ficolin-3 Monoclonal specifically detects Ficolin-3 in Human samples. It is validated for ELISA.
Specifications

Specifications

Ficolin-3
Monoclonal
Alexa Fluor 405
50 mM sodium borate with 0.05% sodium azide
FCN3
Recombinant human H-ficolin
Primary
Clone FCN334 reacts specifically with H-ficolin. No cross-reaction has been observed with L- or M-ficolin
Store at 4°C in the dark.
ELISA
FCN334
ELISA
Collagen/fibrinogen domain-containing lectin 3 p35, Collagen/fibrinogen domain-containing protein 3, FCNHficolin 3, ficolin (collagen/fibrinogen domain containing) 3 (Hakata antigen), ficolin (collagen/fibrinogen domain-containing) 3 (Hakata antigen), ficolin-3, HAKA1MGC22543, Hakata antigen, H-ficolin
Mouse
0.1 mL
8547
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.