Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ficolin-3 Antibody (FCN334) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $560.50
Specifications
Antigen | Ficolin-3 |
---|---|
Clone | FCN334 |
Concentration | 1.0 mg/mL |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Ficolin-3 Monoclonal specifically detects Ficolin-3 in Human samples. It is validated for ELISA, Immunoprecipitation, Sandwich ELISA.Specifications
Ficolin-3 | |
1.0 mg/mL | |
Unconjugated | |
Mouse | |
Human | |
8547 | |
H-Ficolin (Hakata antigen, Ficolin-3) is an innate immunity pattern recognition molecule found in human serum. H-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP) formed by carbohydrates (GlcNAc, GalNAc and fucose) activating complement through the lectin pathway by means of the associated MASPs ((MBL)-associated serine proteases). H-Ficolin consists of 35 kDa protein chains associated in triplet subunits which are again associated in high molecular weight oligomers by means of disulfide bonds at the N-terminal region. | |
Primary | |
Store at 4C in the dark. |
FCN334 | |
Monoclonal | |
Purified | |
RUO | |
Collagen/fibrinogen domain-containing lectin 3 p35, Collagen/fibrinogen domain-containing protein 3, FCNHficolin 3, ficolin (collagen/fibrinogen domain containing) 3 (Hakata antigen), ficolin (collagen/fibrinogen domain-containing) 3 (Hakata antigen), ficolin-3, HAKA1MGC22543, Hakata antigen, H-ficolin | |
FCN3 | |
IgG1 κ | |
Protein A purified | |
Reacts specifically with H-ficolin. No cross-reaction has been observed with L- or M-ficolin. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title