Learn More
Invitrogen™ Ficolin 3 Monoclonal Antibody (FCN334)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ RIG3340102
Description
Reactivity: RIG 334-01-02 reacts with recombinant Ficolin 3 coated directly onto a microtiter well. This antibody can be used as both a capture and detection antibody in sandwich ELISA applications. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
H-Ficolin (Hakata antigen, Ficolin-3) is an innate immunity pattern recognition molecule found in human serum. H-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP) formed by carbohydrates (GlcNAc, GalNAc and fucose) activating complement through the lectin pathway by means of the associated MASPs ((MBL)-associated serine proteases). H-Ficolin consists of 35 kDa protein chains associated in triplet subunits which are again associated in high molecular weight oligomers by means of disulfide bonds at the N-terminal region.
Specifications
Ficolin 3 | |
Monoclonal | |
0.95 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
O75636 | |
FCN3 | |
Recombinant human H-ficolin. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA | |
FCN334 | |
Unconjugated | |
FCN3 | |
Collagen/fibrinogen domain-containing lectin 3 p35; Collagen/fibrinogen domain-containing protein 3; FCN3; FCNH; ficolin (collagen/fibrinogen domain containing) 3 (Hakata antigen); ficolin 3; ficolin-3; HAKA1; Hakata antigen; LOW QUALITY PROTEIN: ficolin-3; MGC22543 | |
Mouse | |
Protein A | |
RUO | |
8547 | |
4°C, store in dark | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.