Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ficolin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571727
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571727 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571727 Supplier Invitrogen™ Supplier No. PA571727
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ficolin 3 Polyclonal Antibody for Western Blot, IHC (P), Flow

H-Ficolin (Hakata antigen, Ficolin-3) is an innate immunity pattern recognition molecule found in human serum. H-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP) formed by carbohydrates (GlcNAc, GalNAc and fucose) activating complement through the lectin pathway by means of the associated MASPs ((MBL)-associated serine proteases). H-Ficolin consists of 35 kDa protein chains associated in triplet subunits which are again associated in high molecular weight oligomers by means of disulfide bonds at the N-terminal region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ficolin 3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FCN3
Gene Accession No. O75636
Gene Alias Collagen/fibrinogen domain-containing lectin 3 p35; Collagen/fibrinogen domain-containing protein 3; FCN3; FCNH; ficolin (collagen/fibrinogen domain containing) 3 (Hakata antigen); ficolin 3; ficolin-3; HAKA1; Hakata antigen; LOW QUALITY PROTEIN: ficolin-3; MGC22543
Gene Symbols FCN3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 214-243 amino acids from the C-terminal region of human FCN3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8547
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.