Learn More
Restek™ FID Base Weldments
SDP
Catalog No. 06611258 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
FID Base Weldment
FID Base Weldment (Capillary Column FID)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06-611-258 FID Base Weldment (Capillary Column FID)
06-611-253 FID Base Weldment
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-611-258 Supplier Restek™ Supplier No. 23053
For Agilent 5890, 6850, or 6890 GCs
  • Meets or exceeds original manufacturer's performance
  • Includes brass nut
  • Available for Agilent 5890, 6850, or 6890 GCs
Specifications
Type FID Base Weldment (Capillary Column FID)
Includes Brass Nut
For Use With (Application) Agilent 6850/6890 GCs
Quantity 1