Learn More
Restek™ FID Collector Housing Kit
SDP
Catalog No. 06611250 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Agilent 5890 FID
Catalog No. For Use With (Equipment)
06-611-250 Agilent 5890 FID
1 options
Catalog No. 06-611-250 Supplier Restek™ Supplier No. 23037
For Agilent 5890 FID
  • Sample pathways deactivated for ultimate inertness
  • Meets or exceeds original manufacturer's performance
    Specifications
    Type FID Collector Housing Kit
    Material Brass
    Quantity 1 Kit
    For Use With (Equipment) Agilent 5890 FID