Learn More
Restek™ FID Jet Removal Tool
SDP
Catalog No. 06611081 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Agilent 5890/6850/6890/7890 FIDs
Catalog No. For Use With (Equipment)
06-611-081 Agilent 5890/6850/6890/7890 FIDs
1 options
Catalog No. 06-611-081 Supplier Restek™ Supplier No. 22328
For Agilent 5890/6850/6890/7890 FIDs
  • Securely grips jet in socket for easy removal or installation
  • Unique, ergonomic handle is easy to hold
Specifications
Type FID Jet Removal Tool
Quantity 1
For Use With (Equipment) Agilent 5890/6850/6890/7890 FIDs