Learn More
Restek™ FID Maintenance Kits
SDP
Catalog No. 06611203 Shop All Restek Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Agilent 5890 GCs
Agilent 6850/6890/7890 GCs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
06-611-203 Agilent 6850/6890/7890 GCs
06-611-204 Agilent 5890 GCs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-611-203 Supplier Restek™ Supplier No. 22179
Only null left
Add to Cart
Add to Cart
For use with Agilent GCs
  • Includes a variety of the most common consumable GC supplies and tools
  • All parts meet or exceed performance of instrument manufacturer's parts
  • Parts list makes reordering easy
Specifications
Type FID Maintenance Kit
Includes Graphite Ferrules (1/4 in., 0.4/0.5/0.8 mm ID), FID/NPD Capillary Adaptor, Capillary Nuts, Jet Reamers/Ferrule Removers, Nut (1/4 in.), Scoring Wafer, Capillary Column Caps, Ignitor, FID Flow-Measuring Adaptor, Wrench (1/4 x 5/16 in.), Installation Gauge, Wire Cleaning Brush, High-Performance Siltek™ Treated FID Dedicated Jet, Spanner Wrench, FID Jet Removal Tool
Quantity 1 Kit
For Use With (Equipment) Agilent 6850/6890/7890 GCs