Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Science First™ Conductive Ink Pen



Click to view available options
:
Conductive ink pen
Description
Includes
Cork board cover; 25 sheets black conductive paper, 20 x 25cm grid; 100 sheets white paper, 20 x 25cm grid; template; conductive ink pen; four connecting wires with alligator clip leads; illustrated instructions
Requires
12VAC power supply or 1.5V battery, digital voltmeter (10 megohms or greater)
- Plot and compare electrical fields by drawing electrodes in conductive ink on semiconducting paper
- Transcribe data to grid paper for a great group lab
- Contains everything needed for a full year's use
- Dimensions: 12.5 x 12 in. (31.8 x 30.5cm)
Specifications
Specifications
Product Type | Conductive Ink Pen |
For Use With (Application) | Cartography training |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction